Assay Details
| Gene Symbol: LOC4330198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771986.2 | XP_015627472.1 | 10 - 11 | 1318 | 73 | |
| XM_015771987.2 | XP_015627473.1 | 9 - 10 | 1256 | 73 | ||
| XM_015771989.2 | XP_015627475.1 | 10 - 11 | 1128 | 73 | ||
| XM_015771990.2 | XP_015627476.1 | 10 - 11 | 1325 | 73 | ||
| GenBank mRNA | FJ765452.1 | - | 9 - 10 | 1234 | 73 | |
More Information
Set Membership: |
|

LOC4330198
Genomic Map