Assay Details
| Gene Symbol: LOC4330198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771986.2 | XP_015627472.1 | 11 - 12 | 1418 | 103 | |
| XM_015771987.2 | XP_015627473.1 | 10 - 11 | 1356 | 103 | ||
| XM_015771989.2 | XP_015627475.1 | 11 - 12 | 1228 | 103 | ||
| GenBank mRNA | AK241507.1 | - | 9 - 10 | 1936 | 103 | |
| FJ765452.1 | - | 10 - 11 | 1334 | 103 | ||
More Information
Set Membership: |
|

LOC4330198
Genomic Map