Assay Details
| Gene Symbol: LOC4330198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771986.2 | XP_015627472.1 | 13 - 14 | 1879 | 78 | |
| XM_015771987.2 | XP_015627473.1 | 12 - 13 | 1817 | 78 | ||
| XM_015771989.2 | XP_015627475.1 | 13 - 14 | 1689 | 78 | ||
| GenBank mRNA | AK241507.1 | - | 11 - 12 | 2397 | 78 | |
| FJ765452.1 | - | 12 - 13 | 1795 | 78 | ||
More Information
Set Membership: |
|

LOC4330198
Genomic Map