Assay Details
| Gene Symbol: LOC4330198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771986.2 | XP_015627472.1 | 2 - 3 | 283 | 134 | |
| XM_015771990.2 | XP_015627476.1 | 2 - 3 | 290 | 134 | ||
More Information
Set Membership: |
|

LOC4330198
Genomic Map