Assay Details
| Gene Symbol: LOC4330198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771986.2 | XP_015627472.1 | 3 - 4 | 448 | 134 | |
| XM_015771987.2 | XP_015627473.1 | 2 - 3 | 386 | 134 | ||
| XM_015771989.2 | XP_015627475.1 | 3 - 4 | 258 | 134 | ||
| XM_015771990.2 | XP_015627476.1 | 3 - 4 | 455 | 134 | ||
| GenBank mRNA | AK241507.1 | - | 2 - 3 | 344 | 134 | |
| FJ765452.1 | - | 2 - 3 | 364 | 134 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771989.2 transcripts.
Set Membership: |
|

LOC4330198
Genomic Map