Assay Details
| Gene Symbol: LOC4330198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771986.2 | XP_015627472.1 | 6 - 7 | 799 | 130 | |
| XM_015771987.2 | XP_015627473.1 | 5 - 6 | 737 | 130 | ||
| XM_015771989.2 | XP_015627475.1 | 6 - 7 | 609 | 130 | ||
| XM_015771990.2 | XP_015627476.1 | 6 - 7 | 806 | 130 | ||
| GenBank mRNA | AK241507.1 | - | 5 - 6 | 695 | 130 | |
| FJ765452.1 | - | 5 - 6 | 715 | 130 | ||
More Information
Set Membership: |
|

LOC4330198
Genomic Map