Assay Details
| Gene Symbol: LOC4330371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770118.2 | XP_015625604.1 | 19 - 20 | 5668 | 68 | |
| XM_015770121.2 | XP_015625607.1 | 6 - 7 | 4207 | 68 | ||
| GenBank mRNA | AK120785.1 | - | 4 - 5 | 2321 | 68 | |
| GQ906768.1 | - | 18 - 19 | 5435 | 68 | ||
More Information
Set Membership: |
|

LOC4330371
Genomic Map