Assay Details
| Gene Symbol: LOC4330421 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767550.2 | XP_015623036.1 | 2 - 3 | 292 | 120 | |
| GenBank mRNA | AK109931.1 | - | 2 - 3 | 292 | 120 | |
More Information
Set Membership: |
|

LOC4330421
Genomic Map