Assay Details
| Gene Symbol: LOC4330987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770018.2 | XP_015625504.1 | 1 - 2 | 281 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770018.2 transcripts.
Set Membership: |
|

LOC4330987
Genomic Map