Assay Details
| Gene Symbol: LOC4330989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768390.2 | XP_015623876.1 | 3 - 4 | 1162 | 157 | |
| XM_015768391.2 | XP_015623877.1 | 4 - 5 | 1374 | 157 | ||
| XM_015768392.2 | XP_015623878.1 | 4 - 5 | 1364 | 157 | ||
| XM_015768393.2 | XP_015623879.1 | 3 - 4 | 1250 | 157 | ||
| GenBank mRNA | AK287814.1 | - | 4 - 5 | 1376 | 157 | |
More Information
Set Membership: |
|

LOC4330989
Genomic Map