Assay Details
| Gene Symbol: LOC4331296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776596.2 | XP_015632082.1 | 23 - 24 | 3690 | 68 | |
| XM_015776597.2 | XP_015632083.1 | 23 - 24 | 3690 | 68 | ||
| XM_015776598.2 | XP_015632084.1 | 23 - 24 | 3675 | 68 | ||
More Information
Set Membership: |
|

LOC4331296
Genomic Map