Assay Details
| Gene Symbol: LOC4332208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777140.2 | XP_015632626.1 | 9 - 10 | 1427 | 96 | |
| XM_015777141.2 | XP_015632627.1 | 9 - 10 | 1336 | 96 | ||
| GenBank mRNA | AK102688.1 | - | 9 - 10 | 1266 | 96 | |
| AK103029.1 | - | 9 - 10 | 1430 | 96 | ||
| AK103791.1 | - | 9 - 10 | 1313 | 96 | ||
More Information
Set Membership: |
|

LOC4332208
Genomic Map