Assay Details
| Gene Symbol: LOC4332367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776930.2 | XP_015632416.1 | 1 - 2 | 461 | 59 | |
| GenBank mRNA | AK071979.1 | - | 1 - 2 | 431 | 59 | |
| AK105349.1 | - | 1 - 2 | 411 | 59 | ||
More Information
Set Membership: |
|

LOC4332367
Genomic Map