Assay Details
| Gene Symbol: LOC4332367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776930.2 | XP_015632416.1 | 5 - 6 | 956 | 97 | |
| GenBank mRNA | AK071979.1 | - | 5 - 6 | 926 | 97 | |
| AK105349.1 | - | 5 - 6 | 906 | 97 | ||
More Information
Set Membership: |
|

LOC4332367
Genomic Map