Assay Details
| Gene Symbol: LOC4332501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777137.2 | XP_015632623.1 | 10 - 11 | 1023 | 101 | |
| XM_015777138.2 | XP_015632624.1 | 10 - 11 | 1023 | 101 | ||
| GenBank mRNA | AK100127.1 | - | 10 - 11 | 1010 | 101 | |
More Information
Set Membership: |
|

LOC4332501
Genomic Map