Assay Details
| Gene Symbol: LOC4333098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772738.2 | XP_015628224.1 | 1 - 2 | 553 | 102 | |
| GenBank mRNA | AK121551.1 | - | 1 - 2 | 544 | 102 | |
| AY302059.1 | - | 1 - 2 | 529 | 102 | ||
More Information
Set Membership: |
|

LOC4333098
Genomic Map