Assay Details
| Gene Symbol: LOC4333271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774707.2 | XP_015630193.1 | 6 - 7 | 855 | 64 | |
| XM_015774708.2 | XP_015630194.1 | 6 - 7 | 855 | 64 | ||
| GenBank mRNA | AK070576.1 | - | 6 - 7 | 862 | 64 | |
More Information
Set Membership: |
|

LOC4333271
Genomic Map