Assay Details
| Gene Symbol: LOC4333271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774707.2 | XP_015630193.1 | 9 - 10 | 1097 | 65 | |
| GenBank mRNA | AK070576.1 | - | 9 - 10 | 1104 | 65 | |
More Information
Set Membership: |
|

LOC4333271
Genomic Map