Assay Details
| Gene Symbol: LOC4333349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776295.2 | XP_015631781.1 | 7 - 8 | 954 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4333349| Gene Symbol: LOC4333349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776295.2 | XP_015631781.1 | 7 - 8 | 954 | 126 | |
Set Membership: |
|