Assay Details
| Gene Symbol: LOC4333393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775840.2 | XP_015631326.1 | 3 - 4 | 859 | 72 | |
| GenBank mRNA | AJ011078.1 | - | 3 - 4 | 837 | 72 | |
| AK061813.1 | - | 3 - 4 | 852 | 72 | ||
| AK098933.1 | - | 3 - 4 | 853 | 72 | ||
| EU267966.1 | - | 3 - 4 | 837 | 72 | ||
| GQ848047.1 | - | 3 - 4 | 837 | 72 | ||
More Information
Set Membership: |
|

LOC4333393
Genomic Map