Assay Details
| Gene Symbol: LOC4334111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775106.2 | XP_015630592.1 | 10 - 11 | 1892 | 70 | |
| GenBank mRNA | AK243482.1 | - | 10 - 11 | 1889 | 70 | |
More Information
Set Membership: |
|

LOC4334111
Genomic Map