Assay Details
| Gene Symbol: LOC4334180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773851.2 | XP_015629337.1 | 2 - 3 | 1362 | 126 | |
| GenBank mRNA | AB099524.1 | - | 2 - 3 | 1001 | 126 | |
| AK063449.1 | - | 2 - 3 | 1362 | 126 | ||
More Information
Set Membership: |
|

LOC4334180
Genomic Map