Assay Details
| Gene Symbol: LOC4334588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772801.2 | XP_015628287.1 | 1 - 2 | 267 | 60 | |
| GenBank mRNA | AK061390.1 | - | 1 - 2 | 159 | 60 | |
| AK104791.1 | - | 1 - 2 | 129 | 60 | ||
More Information
Set Membership: |
|

LOC4334588
Genomic Map