Assay Details
| Gene Symbol: LOC4334618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776908.2 | XP_015632394.1 | 2 - 3 | 642 | 87 | |
| GenBank mRNA | AK058972.1 | - | 2 - 3 | 519 | 87 | |
| AK099193.1 | - | 2 - 3 | 520 | 87 | ||
More Information
Set Membership: |
|

LOC4334618
Genomic Map