Assay Details
| Gene Symbol: LOC4335556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778615.2 | XP_015634101.1 | 4 - 5 | 791 | 86 | |
| GenBank mRNA | AF140495.2 | - | 4 - 5 | 718 | 86 | |
| AK100844.1 | - | 4 - 5 | 721 | 86 | ||
More Information
Set Membership: |
|

LOC4335556
Genomic Map