Assay Details
| Gene Symbol: LOC4336312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781260.2 | XP_015636746.1 | 15 - 16 | 1533 | 116 | |
| GenBank mRNA | AK103837.1 | - | 15 - 16 | 1533 | 116 | |
More Information
Set Membership: |
|

LOC4336312
Genomic Map