Assay Details
| Gene Symbol: LOC4336365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780755.2 | XP_015636241.1 | 1 | 2024 | 70 | |
| GenBank mRNA | AF001505.2 | - | 1 | 1936 | 70 | |
| AK073718.1 | - | 1 | 1953 | 70 | ||
| AK105667.1 | - | 1 | 1677 | 70 | ||
More Information
Set Membership: |
|

LOC4336365
Genomic Map