Assay Details
| Gene Symbol: LOC4336705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781661.2 | XP_015637147.1 | 5 - 6 | 1422 | 62 | |
| GenBank mRNA | AK111914.1 | - | 5 - 6 | 1412 | 62 | |
| AK112099.1 | - | 5 - 6 | 1429 | 62 | ||
More Information
Set Membership: |
|

LOC4336705
Genomic Map