Assay Details
| Gene Symbol: LOC4336705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781661.2 | XP_015637147.1 | 7 - 8 | 1821 | 75 | |
| GenBank mRNA | AB101647.1 | - | 7 - 8 | 1712 | 75 | |
| AK111914.1 | - | 7 - 8 | 1811 | 75 | ||
| AK112099.1 | - | 7 - 8 | 1828 | 75 | ||
More Information
Set Membership: |
|

LOC4336705
Genomic Map