Assay Details
| Gene Symbol: LOC4336705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781661.2 | XP_015637147.1 | 8 - 9 | 2204 | 84 | |
| GenBank mRNA | AB101647.1 | - | 8 - 9 | 2095 | 84 | |
| AK111914.1 | - | 8 - 9 | 2194 | 84 | ||
| AK112099.1 | - | 8 - 9 | 2211 | 84 | ||
More Information
Set Membership: |
|

LOC4336705
Genomic Map