Assay Details
| Gene Symbol: LOC4336715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778882.2 | XP_015634368.1 | 3 - 4 | 563 | 85 | |
| XM_015778883.2 | XP_015634369.1 | 3 - 4 | 674 | 85 | ||
| GenBank mRNA | AK071401.1 | - | 3 - 4 | 486 | 85 | |
More Information
Set Membership: |
|

LOC4336715
Genomic Map