Assay Details
| Gene Symbol: LOC4336724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778102.2 | XP_015633588.1 | 3 - 4 | 410 | 134 | |
| XM_015778103.2 | XP_015633589.1 | 4 - 5 | 482 | 134 | ||
| GenBank mRNA | AK066068.1 | - | 2 - 3 | 511 | 134 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015778102.2, XM_015778103.2 transcripts.
Set Membership: |
|

LOC4336724
Genomic Map