Assay Details
| Gene Symbol: LOC4336724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778102.2 | XP_015633588.1 | 4 - 5 | 539 | 97 | |
| XM_015778103.2 | XP_015633589.1 | 5 - 6 | 611 | 97 | ||
| XM_015778104.2 | XP_015633590.1 | 4 - 5 | 533 | 97 | ||
| GenBank mRNA | AK066068.1 | - | 3 - 4 | 640 | 97 | |
More Information
Set Membership: |
|

LOC4336724
Genomic Map