Assay Details
| Gene Symbol: LOC4336724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778102.2 | XP_015633588.1 | 6 - 7 | 948 | 139 | |
| XM_015778103.2 | XP_015633589.1 | 7 - 8 | 1020 | 139 | ||
| XM_015778104.2 | XP_015633590.1 | 6 - 7 | 942 | 139 | ||
| GenBank mRNA | AK066068.1 | - | 5 - 6 | 1049 | 139 | |
More Information
Set Membership: |
|

LOC4336724
Genomic Map