Assay Details
| Gene Symbol: LOC4337224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780156.2 | XP_015635642.1 | 3 - 4 | 623 | 59 | |
| GenBank mRNA | AK072573.1 | - | 3 - 4 | 630 | 59 | |
| AK109786.1 | - | 3 - 4 | 629 | 59 | ||
More Information
Set Membership: |
|

LOC4337224
Genomic Map