Assay Details
| Gene Symbol: LOC4337224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780156.2 | XP_015635642.1 | 4 - 5 | 894 | 59 | |
| GenBank mRNA | AK072573.1 | - | 4 - 5 | 901 | 59 | |
| AK109786.1 | - | 4 - 5 | 900 | 59 | ||
More Information
Set Membership: |
|

LOC4337224
Genomic Map