Assay Details
| Gene Symbol: LOC4337781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782074.2 | XP_015637560.1 | 3 - 4 | 601 | 136 | |
| XM_015782075.2 | XP_015637561.1 | 3 - 4 | 616 | 136 | ||
| XM_015782076.2 | XP_015637562.1 | 3 - 4 | 616 | 136 | ||
| GenBank mRNA | AK067112.1 | - | 3 - 4 | 602 | 136 | |
More Information
Set Membership: |
|

LOC4337781
Genomic Map