Assay Details
| Gene Symbol: LOC4337781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782074.2 | XP_015637560.1 | 5 - 6 | 759 | 87 | |
| XM_015782075.2 | XP_015637561.1 | 5 - 6 | 774 | 87 | ||
| XM_015782076.2 | XP_015637562.1 | 5 - 6 | 774 | 87 | ||
| GenBank mRNA | AK067112.1 | - | 5 - 6 | 760 | 87 | |
More Information
Set Membership: |
|

LOC4337781
Genomic Map