Assay Details
| Gene Symbol: LOC4337781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782074.2 | XP_015637560.1 | 7 - 8 | 887 | 61 | |
| GenBank mRNA | AK067112.1 | - | 7 - 8 | 888 | 61 | |
More Information
Set Membership: |
|

LOC4337781
Genomic Map