Assay Details
| Gene Symbol: LOC4337781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782074.2 | XP_015637560.1 | 8 - 9 | 951 | 119 | |
| XM_015782075.2 | XP_015637561.1 | 8 - 9 | 963 | 119 | ||
| GenBank mRNA | AK067112.1 | - | 8 - 9 | 952 | 119 | |
More Information
Set Membership: |
|

LOC4337781
Genomic Map