Assay Details
| Gene Symbol: LOC4337784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784614.2 | XP_015640100.1 | 1 - 2 | 575 | 145 | |
| GenBank mRNA | AK068173.1 | - | 1 - 2 | 425 | 145 | |
More Information
Set Membership: |
|

LOC4337784
Genomic Map