Assay Details
| Gene Symbol: LOC4337784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784614.2 | XP_015640100.1 | 2 - 3 | 706 | 74 | |
| GenBank mRNA | AK068173.1 | - | 2 - 3 | 556 | 74 | |
More Information
Set Membership: |
|

LOC4337784
Genomic Map