Assay Details
| Gene Symbol: LOC4337790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784909.2 | XP_015640395.1 | 7 - 8 | 891 | 60 | |
| GenBank mRNA | AK073253.1 | - | 7 - 8 | 812 | 60 | |
More Information
Set Membership: |
|

LOC4337790
Genomic Map