Assay Details
| Gene Symbol: LOC4337790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784909.2 | XP_015640395.1 | 9 - 10 | 1120 | 95 | |
| GenBank mRNA | AK073253.1 | - | 9 - 10 | 1041 | 95 | |
More Information
Set Membership: |
|

LOC4337790
Genomic Map