Assay Details
| Gene Symbol: LOC4337843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782296.2 | XP_015637782.1 | 5 - 6 | 942 | 69 | |
| GenBank mRNA | AK119462.1 | - | 5 - 6 | 951 | 69 | |
More Information
Set Membership: |
|

LOC4337843
Genomic Map