Assay Details
| Gene Symbol: LOC4337843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782296.2 | XP_015637782.1 | 6 - 7 | 1210 | 86 | |
| GenBank mRNA | AK119462.1 | - | 6 - 7 | 1219 | 86 | |
More Information
Set Membership: |
|

LOC4337843
Genomic Map