Assay Details
Gene Symbol: LOC4337855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782038.2 | XP_015637524.1 | 5 - 6 | 654 | 127 | |
XM_015782039.2 | XP_015637525.1 | 3 - 4 | 494 | 127 | ||
GenBank mRNA | AK066581.1 | - | 5 - 6 | 626 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() |
