Assay Details
| Gene Symbol: LOC4339037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784233.2 | XP_015639719.1 | 10 - 11 | 3950 | 135 | |
| XM_015784234.2 | XP_015639720.1 | 10 - 11 | 3947 | 135 | ||
| GenBank mRNA | AK064382.1 | - | 1 - 2 | 856 | 135 | |
More Information
Set Membership: |
|

LOC4339037
Genomic Map