Assay Details
| Gene Symbol: LOC4339037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784233.2 | XP_015639719.1 | 11 - 12 | 4189 | 133 | |
| XM_015784234.2 | XP_015639720.1 | 11 - 12 | 4186 | 133 | ||
| GenBank mRNA | AK064382.1 | - | 2 - 3 | 1095 | 133 | |
More Information
Set Membership: |
|

LOC4339037
Genomic Map