Assay Details
| Gene Symbol: LOC4339037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784233.2 | XP_015639719.1 | 13 - 14 | 4381 | 77 | |
| XM_015784234.2 | XP_015639720.1 | 13 - 14 | 4378 | 77 | ||
| GenBank mRNA | AK064382.1 | - | 4 - 5 | 1287 | 77 | |
More Information
Set Membership: |
|

LOC4339037
Genomic Map